Learn More
New England Biolabs, Inc. BsoBI – 10000 units
SDP
Catalog No. 50812294
Encompass
A restriction endonuclease that recognizes the sequence C^YCGR_G.
Catalog No. 50-812-294 Supplier New England Biolabs, Inc. Supplier No. R0586S
A restriction endonuclease that recognizes the sequence C^YCGR_G.