Learn More
Brady™ BSP™31 Label Attachment System with BBP™33 Label Printer
Catalog No. 19104505 Shop All Brady Corporation Products
Change view
Click to view available options
BSP™31 Label Attachment System with BBP™33 Label Printer
Catalog No.
19-104-505 BSP™31 Label Attachment System with BBP™33 Label Printer
1 options
Catalog No. 19-104-505 Supplier Brady™ Supplier No. BSP31BBP33KIT
Increase the accuracy of specimen tracking and sample management with the BSP™31 Label Attachment System and easy-to-use BBP™33 Label Printer.
  • Print a specimen label on the BBP™33 Printer and apply to a cassette, then simply insert the cassette into the BSP™31 for permanent adhesion
  • Four small heat probes pierce through the corners of the label and cassette and melt the plastic into rivets to permanently hold the label in place
  • Specimen label remains identifiable through histology and anatomic pathology tissue processing protocols and withstands up to 20 years in storage
  • The BSP™31 System must be used exclusively with the B-482 tissue cassette label material and R6400 thermal transfer printer ribbon from Brady™
Specifications
Product Type Printer Kit
Autoclavable Not Autoclavable
Description Printer Kit
Includes B33-141-481 slide labels; B33-199-482 tissue cassette labels; B30-R6400 print ribbon
Shape Rectangle
Quantity Kit