Learn More
New England Biolabs, Inc. Bsp1286I – 500 units
SDP
Catalog No. 50811961
Encompass
A restriction endonuclease that recognizes the sequence G_DGCH^C.
Catalog No. 50-811-961 Supplier New England Biolabs, Inc. Supplier No. R0120S
A restriction endonuclease that recognizes the sequence G_DGCH^C.