Learn More
New England Biolabs, Inc. BspCNI – 100 units
SDP
Catalog No. 50812366
Encompass
A restriction endonuclease that recognizes the sequence CTCAGNNNNNNN_NN^.
Catalog No. 50-812-366 Supplier New England Biolabs, Inc. Supplier No. R0624S
A restriction endonuclease that recognizes the sequence CTCAGNNNNNNN_NN^.