Learn More
New England Biolabs, Inc. BspDI – 2000 units
SDP
Catalog No. 50812239
Encompass
A restriction endonuclease that recognizes the sequence AT^CG_AT.
Catalog No. 50-812-239 Supplier New England Biolabs, Inc. Supplier No. R0557S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence AT^CG_AT.