Learn More
New England Biolabs, Inc. BspEI – 5000 units
SDP
Catalog No. 50812210
Encompass
A restriction endonuclease that recognizes the sequence T^CCGG_A.
Catalog No. 50-812-210 Supplier New England Biolabs, Inc. Supplier No. R0540L
A restriction endonuclease that recognizes the sequence T^CCGG_A.