Learn More
New England Biolabs, Inc. BspHI – 2500 units
SDP
Catalog No. 50812165
Encompass
A restriction endonuclease that recognizes the sequence T^CATG_A.
Catalog No. 50-812-165 Supplier New England Biolabs, Inc. Supplier No. R0517L
A restriction endonuclease that recognizes the sequence T^CATG_A.