Learn More
New England Biolabs, Inc. BspHI – 500 units
SDP
Catalog No. 50812166
Encompass
A restriction endonuclease that recognizes the sequence T^CATG_A.
Catalog No. 50-812-166 Supplier New England Biolabs, Inc. Supplier No. R0517S
A restriction endonuclease that recognizes the sequence T^CATG_A.