Learn More
New England Biolabs, Inc. BspMI – 100 units
SDP
Catalog No. 50812142
Encompass
A restriction endonuclease that recognizes the sequence ACCTGCNNNN^NNNN_.
Catalog No. 50-812-142 Supplier New England Biolabs, Inc. Supplier No. R0502S
A restriction endonuclease that recognizes the sequence ACCTGCNNNN^NNNN_.