Learn More
New England Biolabs, Inc. BspQI – 2500 units
SDP
Catalog No. 50994944
Encompass
A restriction endonuclease that recognizes the sequence GCTCTTCN^NNN_.
Catalog No. 50-994-944 Supplier New England Biolabs, Inc. Supplier No. R0712L
A restriction endonuclease that recognizes the sequence GCTCTTCN^NNN_.