Learn More
New England Biolabs, Inc. BspQI – 500 units
SDP
Catalog No. 50994933
Encompass
A restriction endonuclease that recognizes the sequence GCTCTTCN^NNN_.
Catalog No. 50-994-933 Supplier New England Biolabs, Inc. Supplier No. R0712S
A restriction endonuclease that recognizes the sequence GCTCTTCN^NNN_.