Learn More
New England Biolabs, Inc. BspQI HF-2500 units
SDP
Catalog No. NC3594300
Encompass
A Type IIS restriction endonuclease that recognizes the sequence GCTCTTC(1/4). An E. coli strain that carries the modified cloned BspQI gene from Bacillus sphaericus. BspQI-HF is an engineered and improved version of BspQI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)
Catalog No. NC3594300 Supplier New England Biolabs, Inc. Supplier No. R3712L
A Type IIS restriction endonuclease that recognizes the sequence GCTCTTC(1/4). An E. coli strain that carries the modified cloned BspQI gene from Bacillus sphaericus. BspQI-HF is an engineered and improved version of BspQI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)