Learn More
New England Biolabs, Inc. BspQI HF-500 units
SDP
Catalog No. 503025699
Encompass
A Type IIS restriction endonuclease that recognizes the sequence GCTCTTC(1/4). An E. coli strain that carries the modified cloned BspQI gene from Bacillus sphaericus. BspQI-HF is an engineered and improved version of BspQI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)
Catalog No. 50-302-5699 Supplier New England Biolabs, Inc. Supplier No. R3712S
A Type IIS restriction endonuclease that recognizes the sequence GCTCTTC(1/4). An E. coli strain that carries the modified cloned BspQI gene from Bacillus sphaericus. BspQI-HF is an engineered and improved version of BspQI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)