Learn More
New England Biolabs, Inc. BsrBI – 5000 units
SDP
Catalog No. 50811922
Encompass
A restriction endonuclease that recognizes the sequence CCG_^CTC.
Catalog No. 50-811-922 Supplier New England Biolabs, Inc. Supplier No. R0102L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
A restriction endonuclease that recognizes the sequence CCG_^CTC.