Learn More
New England Biolabs, Inc. BsrDI – 200 units
SDP
Catalog No. 50812272
Encompass
A restriction endonuclease that recognizes the sequence GCAATG_NN^.
Catalog No. 50-812-272 Supplier New England Biolabs, Inc. Supplier No. R0574S
A restriction endonuclease that recognizes the sequence GCAATG_NN^.