Learn More
New England Biolabs, Inc. BsrFI-v2 – 1000 units
SDP
Catalog No. 501526445
Encompass
A restriction endonuclease that recognizes the sequence R^CCGG_Y.
Catalog No. 50-152-6445 Supplier New England Biolabs, Inc. Supplier No. R0682S
A restriction endonuclease that recognizes the sequence R^CCGG_Y.