Learn More
New England Biolabs, Inc. BsrI – 1000 units
SDP
Catalog No. 50812186
Encompass
A restriction endonuclease that recognizes the sequence ACTG_GN^.
Catalog No. 50-812-186 Supplier New England Biolabs, Inc. Supplier No. R0527S
A restriction endonuclease that recognizes the sequence ACTG_GN^.