Learn More
New England Biolabs, Inc. BsrI – 5000 units
SDP
Catalog No. 50812185
Encompass
A restriction endonuclease that recognizes the sequence ACTG_GN^.
Catalog No. 50-812-185 Supplier New England Biolabs, Inc. Supplier No. R0527L
A restriction endonuclease that recognizes the sequence ACTG_GN^.