Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medex Supply BSS PLUS Sterile Intraocular Irrigating Solution, 500 mL, preservative-free, sterile
SDP

Catalog No. NC2599139
Encompass

BSS PLUS Sterile Intraocular Irrigating Solution is a sterile and preservative-free solution expertly designed for intraocular surgical procedures. It effectively mimics the natural composition of aqueous humor, offering optimal physiological support during operations. Its advanced formulation emphasizes maintaining corneal integrity and enhancing surgical outcomes. This solution comes in a two-part system, requiring aseptic reconstitution for guaranteed stability before use.

  • Volume: 500 mL total reconstituted
  • Sterile and preservative-free solution
  • Designed for intraocular irrigation during ophthalmic surgery
  • Enriched with bicarbonate, dextrose, and glutathione
  • Maintains corneal endothelial integrity
  • Reconstitution just prior to use
  • Discard time post-reconstitution: 6 hours
  • Storage temperature: 2 to 25 °C (36 to 77 °F)
  • Not for injection or intravenous infusion
  • Single use only

Catalog No. NC2599139 Supplier Medex Supply Supplier No. 0065080050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BSS PLUS Sterile Intraocular Irrigating Solution is a sterile and preservative-free solution expertly designed for intraocular surgical procedures. It effectively mimics the natural composition of aqueous humor, offering optimal physiological support during operations. Its advanced formulation emphasizes maintaining corneal integrity and enhancing surgical outcomes. This solution comes in a two-part system, requiring aseptic reconstitution for guaranteed stability before use.

  • Volume: 500 mL total reconstituted
  • Sterile and preservative-free solution
  • Designed for intraocular irrigation during ophthalmic surgery
  • Enriched with bicarbonate, dextrose, and glutathione
  • Maintains corneal endothelial integrity
  • Reconstitution just prior to use
  • Discard time post-reconstitution: 6 hours
  • Storage temperature: 2 to 25 °C (36 to 77 °F)
  • Not for injection or intravenous infusion
  • Single use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.