Learn More
New England Biolabs, Inc. BssHII (25,000 units/ml) – 2500 units
SDP
Catalog No. 50812135
Encompass
A restriction endonuclease that recognizes the sequence G^CGCG_C.
Catalog No. 50-812-135 Supplier New England Biolabs, Inc. Supplier No. R0199M
A restriction endonuclease that recognizes the sequence G^CGCG_C.