Learn More
New England Biolabs, Inc. BssHII – 500 units
SDP
Catalog No. 50812136
Encompass
A restriction endonuclease that recognizes the sequence G^CGCG_C.
Catalog No. 50-812-136 Supplier New England Biolabs, Inc. Supplier No. R0199S
A restriction endonuclease that recognizes the sequence G^CGCG_C.