Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse BSSP 4 (PRSS22) ELISA Kit

Catalog No. EMPRSS22X10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMPRSS22X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EMPRSS22X10 Supplier Invitrogen™ Supplier No. EMPRSS22X10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

The Mouse BSSP 4 (PRSS22) ELISA quantitates Ms BSSP 4 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms BSSP 4. Principle of the method The Mouse BSSP 4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9ER10
Assay Range 60-4000 pg/mL
Assay Sensitivity 60 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Proteases Peptidases & Inhibitors
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 4733401N09Rik; brain-specific serine protease 4; BSP-2; BSSP4; BSSP-4; hBSSP-4; MGC9599; prosemin; protease, serine 22; protease, serine S1 family member 22; protease, serine, 22; PRSS22; Prss26; Serine protease 22; serine protease 26; SP001LA; Tryptase epsilon; UNQ302/PRO3434733401N09Rik; brain-specific serine protease 4; BSP-2; BSSP4; BSSP-4; hBSSP-4; MGC9599; prosemin; protease, serine 22; protease, serine S1 family member 22; protease, serine, 22; PRSS22; Prss26; Serine protease 22; serine protease 26; SP001LA; Tryptase epsilon; UNQ302/PRO343; 4733401N09Rik; BSSP-4; SP001LA
Gene ID (Entrez) 70835
Gene Symbol PRSS22
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases BSSP-4, Serine protease 22, Serine protease 26, Tryptase epsilon
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target BSSP4
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.