Learn More
New England Biolabs, Inc. BssSI-v2 – 1000 units
SDP
Catalog No. 501047491
Encompass
BssSI-v2 is an engineered and improved version of BssSI that recognizes the sequence VC^TCGAGB. BssSI-v2 is an engineered and improved version of BssSI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)
Catalog No. 50-104-7491 Supplier New England Biolabs, Inc. Supplier No. R0680L
BssSI-v2 is an engineered and improved version of BssSI that recognizes the sequence VC^TCGAGB. BssSI-v2 is an engineered and improved version of BssSI. (NEB uses the designation "v2" or "HF" in the name to indicate engineered enzymes.)