Learn More
New England Biolabs, Inc. Bst DNA Polymerase, Full Length – 500 units
SDP
Catalog No. 504367
Encompass
Bst DNA Polymerase, Full Length is the full length polymerase from Bacillus stearothermophilus. It has 5' to 3' polymerase activity and double-strand specific 5' to 3' exonuclease activity, but lacks 3' to 5' exonuclease activity.
Catalog No. 50-436-7 Supplier New England Biolabs, Inc. Supplier No. M0328S
Bst DNA Polymerase, Full Length is the full length polymerase from Bacillus stearothermophilus. It has 5' to 3' polymerase activity and double-strand specific 5' to 3' exonuclease activity, but lacks 3' to 5' exonuclease activity.