Learn More
New England Biolabs, Inc. Bst DNA Polymerase, Large Fragment – 1600 units
SDP
Catalog No. 50811704
Encompass
Bst DNA Polymerase, Large Fragment is the portion of the Bacillus stearothermophilus DNA Polymerase protein that contains the 5' to 3' polymerase activity, but lacks 5' to 3' exonuclease activity.
Catalog No. 50-811-704 Supplier New England Biolabs, Inc. Supplier No. M0275S
Bst DNA Polymerase, Large Fragment is the portion of the Bacillus stearothermophilus DNA Polymerase protein that contains the 5' to 3' polymerase activity, but lacks 5' to 3' exonuclease activity.