Learn More
New England Biolabs, Inc. Bst DNA Polymerase, Large Fragment – 8000 units
SDP
Supplier:  New England Biolabs, Inc. M0275M
Encompass
Bst DNA Polymerase, Large Fragment is the portion of the Bacillus stearothermophilus DNA Polymerase protein that contains the 5' to 3' polymerase activity, but lacks 5' to 3' exonuclease activity.
Catalog No. 50-811-703