Learn More
New England Biolabs, Inc. Bst DNA Polymerase, Large Fragment – 8000 units
SDP
Catalog No. 50811702
Encompass
Bst DNA Polymerase, Large Fragment is the portion of the Bacillus stearothermophilus DNA Polymerase protein that contains the 5' to 3' polymerase activity, but lacks 5' to 3' exonuclease activity.
Catalog No. 50-811-702 Supplier New England Biolabs, Inc. Supplier No. M0275L
Bst DNA Polymerase, Large Fragment is the portion of the Bacillus stearothermophilus DNA Polymerase protein that contains the 5' to 3' polymerase activity, but lacks 5' to 3' exonuclease activity.