Learn More
Meridian Life Science Bst DNA Polymerase, 10ML
Supplier: Meridian Life Science MDX01210
Bst DNA Polymerase is an enzyme derived from the large fragment of Bacillus stearothermophilus DNA Polymerase I. It contains 5´- 3´ DNA polymerase activity and strong strand displacement activity but lacks 5´- 3´ exonuclease activity. The strong strand displacement activity enables Bst DNA Polymerase to synthesize DNA at a constant temperature making it an ideal enzyme for isothermal amplification, including HDA, MCA, and Loop-Mediated Isothermal DNA Amplification (LAMP). It is supplied with an Enzyme Dilution Buffer and a Bst Reaction Buffer formulated to deliver the fastest amplification speed (time-to-results), yield, salt tolerance and sensitivity, speeding up time for optimization. For the most difficult templates, an Inhibitor-Tolerant Bst Buffer is available that is specifically designed for high performance when using crude samples such as sputum or saliva.