Learn More
New England Biolabs, Inc. Bst 2.0 DNA Polymerase – 8000 units
SDP
Catalog No. NC0328649
Encompass
Bst 2.0 DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment (Bst DNA Polymerase, Large Fragment). Bst 2.0 DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand displacement activity but lacks 5' to 3' exonuclease activity. Bst 2.0 DNA Polymerase displays improved amplification speed, yield, salt tolerance and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.
Catalog No. NC0328649 Supplier New England Biolabs, Inc. Supplier No. M0537L
Bst 2.0 DNA Polymerase is an in silico designed homologue of Bacillus stearothermophilus DNA Polymerase I, Large Fragment (Bst DNA Polymerase, Large Fragment). Bst 2.0 DNA Polymerase contains 5' to 3' DNA polymerase activity and strong strand displacement activity but lacks 5' to 3' exonuclease activity. Bst 2.0 DNA Polymerase displays improved amplification speed, yield, salt tolerance and thermostability compared to wild-type Bst DNA Polymerase, Large Fragment.