Learn More
Meridian Life Science Bst Reaction Buffer 10x, 5ML
SDP
Catalog No. 5000856407
Encompass_Preferred
Bst Reaction Buffer, 10x is optimized for use with Bst DNA Polymerase (MDX012 or MDX018), it formulated to deliver the fastest amplification speed (time-to-results), yield, salt tolerance and sensitivity, speeding up time for optimization.
Catalog No. 50-008-56407 Supplier Meridian Life Science Supplier No. MDX0765
Bst Reaction Buffer, 10x is optimized for use with Bst DNA Polymerase (MDX012 or MDX018), it formulated to deliver the fastest amplification speed (time-to-results), yield, salt tolerance and sensitivity, speeding up time for optimization.