Learn More
New England Biolabs, Inc. Bst-XT WarmStart™ DNA Polymerase - 1600 units
SDP
Catalog No. 5000077830
Encompass
Bst-XT WarmStart™ DNA Polymerase is an engineered variant of Bacillus stearothermophilus DNA Polymerase, Large Fragment fused to a novel nucleic acid binding domain for improved isothermal amplification performance and increased reverse transcription activity. This enzyme combines the polymerization speed of Bst 3.0 and the high specificity of Bst 2.0 DNA polymerases. Bst-XT WarmStart DNA Polymerase contains a reversibly bound aptamer, which inhibits polymerase activity at room temperature. The aptamer rapidly releases above 45°C, therefore no separate activation step is needed. Bst-XT WarmStart DNA Polymerase contains 5' to 3' DNA polymerase activity with either DNA or RNA templates and strong strand displacement activity, but lacks 5' to 3' and 3' to 5' exonuclease activity.
Catalog No. 50-000-77830 Supplier New England Biolabs, Inc. Supplier No. M9204S
Bst-XT WarmStart™ DNA Polymerase is an engineered variant of Bacillus stearothermophilus DNA Polymerase, Large Fragment fused to a novel nucleic acid binding domain for improved isothermal amplification performance and increased reverse transcription activity. This enzyme combines the polymerization speed of Bst 3.0 and the high specificity of Bst 2.0 DNA polymerases. Bst-XT WarmStart DNA Polymerase contains a reversibly bound aptamer, which inhibits polymerase activity at room temperature. The aptamer rapidly releases above 45°C, therefore no separate activation step is needed. Bst-XT WarmStart DNA Polymerase contains 5' to 3' DNA polymerase activity with either DNA or RNA templates and strong strand displacement activity, but lacks 5' to 3' and 3' to 5' exonuclease activity.