Learn More
New England Biolabs, Inc. Bst-XT WarmStart™ DNA Polymerase (Glycerol-free) - 8000 units
SDP
Supplier:  New England Biolabs, Inc. M9205L
Encompass
Bst-XT WarmStart™ DNA Polymerase (Glycerol-free) is formulated without glycerol to support lyophilization, incorporation into microfluidic devices, and enable quick adoption into automation workflows. Bst-XT WarmStart DNA Polymerase is an engineered variant of Bacillus stearothermophilus DNA Polymerase, Large Fragment fused to a novel nucleic acid binding domain for improved isothermal amplification performance and increased reverse transcription activity. This enzyme combines the polymerization speed of Bst 3.0 and the high specificity of Bst 2.0 DNA polymerases. Bst-XT WarmStart DNA Polymerase contains a reversibly bound aptamer, which inhibits polymerase activity at room temperature. The aptamer rapidly releases above 45°C, therefore no separate activation step is needed. Bst-XT WarmStart DNA Polymerase contains 5' to 3' DNA polymerase activity with either DNA or RNA templates and strong strand displacement activity, but lacks 5' to 3' and 3' to 5' exonuclease activity.
Catalog No. 50-000-77831