Learn More
New England Biolabs, Inc. BstAPI – 1000 units
SDP
Catalog No. NC9844398
Encompass
A restriction endonuclease that recognizes the sequence GCAN_NNN^NTGC.
Catalog No. NC9844398 Supplier New England Biolabs, Inc. Supplier No. R0654L
A restriction endonuclease that recognizes the sequence GCAN_NNN^NTGC.