Learn More
New England Biolabs, Inc. BstAPI – 200 units
SDP
Catalog No. 504462
Encompass
A restriction endonuclease that recognizes the sequence GCAN_NNN^NTGC.
Catalog No. 50-446-2 Supplier New England Biolabs, Inc. Supplier No. R0654S
A restriction endonuclease that recognizes the sequence GCAN_NNN^NTGC.