Learn More
New England Biolabs, Inc. BstBI – 12500 units
SDP
Catalog No. 50812169
Encompass
A restriction endonuclease that recognizes the sequence TT^CG_AA.
Catalog No. 50-812-169 Supplier New England Biolabs, Inc. Supplier No. R0519L
A restriction endonuclease that recognizes the sequence TT^CG_AA.