Learn More
New England Biolabs, Inc. BstEII – 2000 units
SDP
Catalog No. 50812067
Encompass
A restriction endonuclease that recognizes the sequence G^GTNAC_C.
This item is not returnable. View return policy
Catalog No. 50-812-067 Supplier New England Biolabs, Inc. Supplier No. R0162S
A restriction endonuclease that recognizes the sequence G^GTNAC_C.