Learn More
Selleck Chemical LLC BSTFA-TMCS S5443-25mg
SDP
Catalog No. 503110341
Encompass_Preferred
BSTFA-TMCS is used as a derivatizing agent in chromatography It functions as a silylation reagent without additional solvents The combination of BSTFA and TMCS is the preferred reagent for trimethylsilylation of alcohols alkaloids amines biogenic amines carboxylic acids phenols and steroids
Catalog No. 50-311-0341 Supplier Selleck Chemical LLC Supplier No. S544325MG
BSTFA-TMCS is used as a derivatizing agent in chromatography It functions as a silylation reagent without additional solvents The combination of BSTFA and TMCS is the preferred reagent for trimethylsilylation of alcohols alkaloids amines biogenic amines carboxylic acids phenols and steroids