Learn More
New England Biolabs, Inc. BstNI – 3000 units
SDP
Catalog No. 50812075
Encompass
A restriction endonuclease that recognizes the sequence CC^W_GG.
Catalog No. 50-812-075 Supplier New England Biolabs, Inc. Supplier No. R0168S
A restriction endonuclease that recognizes the sequence CC^W_GG.