Learn More
New England Biolabs, Inc. BstUI – 1000 units
SDP
Catalog No. 50812168
Encompass
A restriction endonuclease that recognizes the sequence CG_^CG.
Catalog No. 50-812-168 Supplier New England Biolabs, Inc. Supplier No. R0518S
A restriction endonuclease that recognizes the sequence CG_^CG.