Learn More
New England Biolabs, Inc. BstUI – 5000 units
SDP
Catalog No. 50812167
Encompass
A restriction endonuclease that recognizes the sequence CG_^CG.
Catalog No. 50-812-167 Supplier New England Biolabs, Inc. Supplier No. R0518L
A restriction endonuclease that recognizes the sequence CG_^CG.