Learn More
New England Biolabs, Inc. BstXI – 1000 units
SDP
Catalog No. 50811949
Encompass
A restriction endonuclease that recognizes the sequence CCAN_NNNN^NTGG.
Catalog No. 50-811-949 Supplier New England Biolabs, Inc. Supplier No. R0113S
A restriction endonuclease that recognizes the sequence CCAN_NNNN^NTGG.