Learn More
New England Biolabs, Inc. Bsu DNA Polymerase, Large Fragment – 200 units
SDP
Catalog No. 504369
Encompass
Bsu DNA Polymerase I, Large Fragment retains the 5' to 3' polymerase activity of the Bacillus subtilis DNA polymerase I, but lacks the 5' to 3' exonuclease domain. This large fragment naturally lacks 3' to 5' exonuclease activity.
Catalog No. 50-436-9 Supplier New England Biolabs, Inc. Supplier No. M0330S
Bsu DNA Polymerase I, Large Fragment retains the 5' to 3' polymerase activity of the Bacillus subtilis DNA polymerase I, but lacks the 5' to 3' exonuclease domain. This large fragment naturally lacks 3' to 5' exonuclease activity.