Learn More
INTACT GENOMICS INC Bsu DNA Polymerase Large Fragment 1000 Units (5 units/µl)
Catalog No. 5000082925
Encompass
Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification). Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity (1)Applications• Strand displacement DNA synthesis (2)• Random primer labeling• Second strand cDNA synthesis• dA-tailing
Catalog No. 50-000-82925 Supplier INTACT GENOMICS INC Supplier No. 3585
Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification). Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity (1)Applications• Strand displacement DNA synthesis (2)• Random primer labeling• Second strand cDNA synthesis• dA-tailing