Learn More
INTACT GENOMICS INC Bsu DNA Polymerase Large Fragment 200 Units (5 units/µl)
Catalog No. 5000082924
Encompass
Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification). Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity (1)Applications• Strand displacement DNA synthesis (2)• Random primer labeling• Second strand cDNA synthesis• dA-tailing
Catalog No. 50-000-82924 Supplier INTACT GENOMICS INC Supplier No. 3582
Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification). Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity (1)Applications• Strand displacement DNA synthesis (2)• Random primer labeling• Second strand cDNA synthesis• dA-tailing