Learn More
INTACT GENOMICS INC Bsu DNA Polymerase Large Fragment, Lyophilized 2000 Units (5 units/µl)
Catalog No. 5000082825
Encompass
Lyophilized Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification).Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.
Catalog No. 50-000-82825 Supplier INTACT GENOMICS INC Supplier No. 3587LYO
Lyophilized Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification).Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.