Learn More
INTACT GENOMICS INC Bsu DNA Polymerase Large Fragment, Lyophilized 200 Units (5 units/µl)
Catalog No. 5000082816
Encompass
Lyophilized Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification).Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.
Catalog No. 50-000-82816 Supplier INTACT GENOMICS INC Supplier No. 3582LYO
Lyophilized Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification).Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.