Learn More
INTACT GENOMICS INC Bsu DNA Polymerase Large Fragment, Glycerol Free 1000 Units (5 units/µl)
Catalog No. 5000083028
Encompass
Glycerol Free or Lyophilized Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification).Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.
Catalog No. 50-000-83028 Supplier INTACT GENOMICS INC Supplier No. 3585GF
Glycerol Free or Lyophilized Bsu DNA Polymerase I, Large Fragment is suitable for various purposes including RPA (Recombinase Polymerase Amplification).Bsu DNA Polymerase I, Large Fragment is a product of the Bacillus subtilis DNA polymerase I which lacks the N-terminal exonuclease domain (1-296 amino acids). It retains the 5´→ 3´ polymerase activity of DNA polymerase I but lacks the 5´→ 3´ exonuclease activity. This large fragment also lacks 3´→ 5´ exonuclease activity.