Learn More
New England Biolabs, Inc. Bsu36I – 5000 units
SDP
Catalog No. 50812177
Encompass
A restriction endonuclease that recognizes the sequence CC^TNA_GG.
Catalog No. 50-812-177 Supplier New England Biolabs, Inc. Supplier No. R0524L
A restriction endonuclease that recognizes the sequence CC^TNA_GG.