Learn More
Bon Opus Biosciences Biotinylated Human Carbonic Anhydrase 9 (C-Avi-6His) -20ug
SDP
Supplier:  Bon Opus Biosciences CY0820UG
Encompass_Preferred
Biotinylated Recombinant Human Carbonic Anhydrase 9 is produced by our Mammalian expression system and the target gene encoding Gln38-Asp414 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# Q16790. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY08
Catalog No. 50-208-0490