Learn More
Bon Opus Biosciences Biotinylated Human EGFR vIII (C-6His-Avi) -100ug
SDP
Supplier:  Bon Opus Biosciences CY02100UG
Encompass_Preferred
Biotinylated Recombinant Human Epidermal Growth Factor Receptor/Receptor Tyrosine Protein KinaseErbB1 is produced by our Mammalian expression system and the target gene encoding Leu25-Ser378 is expressed with a 6His, Avi tag at the C-terminus. Expression system: Human Cells. Accession# NP_001333870. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# CY02
Catalog No. 50-208-0480